Learn More
Accuform™ STOPOUT™ Gate Valve Lockout
Lockout devices control energy sources and ensure machine and equipment remain inoperable to prevent unexpected start-up or release of stored energy in accordance with OSHA 29 CFR 1910.147 requirements
$24.39 - $36.92
| Numéro de catalogue | Numéro du manufacturier. | Diameter (English) | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Diameter (English) | Prix | Quantité | |||||
|
19211290
|
Accuform™
KDD470 |
1 to 2-1/2 in. |
chaque for $24.39
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
19211291
|
Accuform™
KDD471 |
2-1/2 to 5 in. |
chaque for $36.92
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
- Hinged clamshell housing is designed to fully enclose the valve handle to prevent unauthorized access
- Tough thermoplastic, polypropylene device is a bright hazard red color, and features a durable, impact-resistant aluminum hinge with permanent binding pin
- Includes DANGER LOCKED OUT labels (English, Spanish, and French) with space to write more information as needed
- Red octagon shape reinforces lockout status and conveys STOP
Spécifications
| Gate Valve Lockout | |
| Plastic |