missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spiking Solution for Water and Soil - CLP, Ricca Chemical
For Water and Soil
Supplier: Ricca Chemical Company RSPIKE1100
Specifications
| Spiking Solution | |
| 7732-18-5 , 7697-37-2 , 7784-27-2 , 10022-31-8 , 7782-49-2 , 7440-28-0 | |
| 95.0,6.8,0.19,0.03,0.01,0.01 | |
| Colorless | |
| Approximately 0°C | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| Calibration Standard | |
| CLP |
| Amber Poly Bottle | |
| 91.0,6.53,0.18,0.03,0.01,0.01 | |
| Liquid | |
| Acidic | |
| Approximately 100°C | |
| Miscible with water | |
| 100 mL |