CAS RN 7440-28-0
Thallium rod, 12.7mm (0.5in) dia, 99.99% (metals basis)
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium rod, 12.7mm (0.5in) dia, 99.999% (metals basis)
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N Synonym: ramor,thallium, metallic,thallium, elemental,unii-ad84r52xlf,thallium, soluble compounds,ad84r52xlf,thallium, elemental and soluble compounds,rod, 12.7mm 0.5in dia,granules,thallium PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium ICP Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium AA Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium ICP-MS Standard, 1000 ppm Tl in 3% HNO3, Ricca Chemical
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium ICP Standard, 10,000 ppm Tl in 5% HNO3, Ricca Chemical
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]
Thallium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-28-0 Molecular Formula: Tl Molecular Weight (g/mol): 204.38 MDL Number: MFCD00134063 InChI Key: BKVIYDNLLOSFOA-UHFFFAOYSA-N PubChem CID: 5359464 ChEBI: CHEBI:30440 IUPAC Name: thallium SMILES: [Tl]