CAS RN 7782-49-2
Selenium, 99.5+%, powder, -200 mesh
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium standard solution, for AAS, 1 mg/ml Se in 10% HCl
CAS: 7782-49-2 | Se | 78.97 g/mol
Selenium, 99+%, powder
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium, AAS standard solution, Specpure™ Se 1000μg/mL
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium shot, amorphous, 2-6mm (0.08-0.2in), Puratronic™, 99.999% (metals basis)
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium shot, amorphous, 1-6mm (0.04-0.24in), Puratronic, 99.999+% (metals basis)
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium powder, -200 mesh, 99.999% (metals basis)
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Kjeldahl tablets with selenium (Wieninger)
CAS: 7782-49-2
Selenium ingot/button,≈38mm (1.5 in.) dia.
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N Synonym: selenium, elemental,selen,hydride,atom,elemental,selenio,vandex,alloy,and compounds,gray PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium ICP-MS Standard, 1000 ppm Se in 3% HNO3, Ricca Chemical
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Se), LabChem™
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium ICP Standard, 10,000 ppm Se in 5% HNO3, Ricca Chemical
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.971 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium ICP Standard, 1000 ppm Se in 3% HNO3, Ricca Chemical
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
Selenium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7782-49-2 Molecular Formula: Se Molecular Weight (g/mol): 78.97 MDL Number: MFCD00134090 MFCD00011224 InChI Key: BUGBHKTXTAQXES-UHFFFAOYSA-N PubChem CID: 6326970 ChEBI: CHEBI:27568 IUPAC Name: selenium SMILES: [Se]
- 1
- 2