CAS RN 10022-31-8
Barium nitrate, ACS, 99+%
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium nitrate, 99.95% (metals basis)
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium nitrate, Puratronic™, 99.999% (metals basis)
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium nitrate, 99+%, for analysis
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium, plasma standard solution, Specpure™ Ba 1000μg/mL
CAS: 10022-31-8 Molecular Weight (g/mol): 261.34 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium nitrate, ACS reagent
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium nitrate, 99%
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N Synonym: barium nitrate,nitrobarite,nitric acid, barium salt,barium dinitrate,bariumnitrate,dusicnan barnaty czech,nitrato barico spanish,unii-mdc5sw56xc,nitrate de baryum french,barium nitrate ba no3 2 PubChem CID: 24798 IUPAC Name: barium(2+);dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium ICP Standard, 1000 ppm Ba in 3% HNO3, Ricca Chemical
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium ICP Standard, 10,000 ppm Ba in 5% HNO3, Ricca Chemical
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Barium ICP-MS Standard, 1000 ppm Ba in 3% HNO3, Ricca Chemical
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Barium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
VeriSpec™ Barium Standard for ICP/MS 100 ppm in 2% HNO3, Ricca Chemical
CAS: 10022-31-8 Molecular Formula: BaN2O6 Molecular Weight (g/mol): 261.34 MDL Number: MFCD00003442 InChI Key: IWOUKMZUPDVPGQ-UHFFFAOYSA-N PubChem CID: 24798 IUPAC Name: barium(2+) dinitrate SMILES: [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
- 1
- 2