missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sphingosine-1-phosphate

Supplier:  Thermo Scientific Chemicals J66459LB0

Quantity 5 mg

 Afficher plus de versions de ce produit

Numéro de catalogue. AAJ66459LB0

Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

26993-30-6
C18H38NO5P
D-erythro-Sphingosine 1-phosphate; S1P

Spécifications

5 mg
-30°C to -10°C
C18H38NO5P
D-erythro-Sphingosine 1-phosphate; S1P
Sphingosine-1-phosphate
DUYSYHSSBDVJSM-KRWOKUGFSA-N
{[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
White to off-white
26993-30-6
379.47
Light Sensitive
Powder
CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
379.48
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Store at -20°C; Store under Nitrogen

FDS
Documents

Documents

Promotions

Promotions