missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sphingosine-1-phosphate

Catalog No. p-7070105
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66459MCR 1 mg
AAJ66459LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66459MCR Supplier Thermo Scientific Chemicals Supplier No. J66459MCR
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26993-30-6
Molecular Formula C18H38NO5P
Synonym D-erythro-Sphingosine 1-phosphate; S1P

Specifications

Quantity 1 mg
Description White to off-white
Content And Storage -30°C to -10°C
Formula Weight 379.47
Sensitivity Light Sensitive
Product Type Sphingosine-1-phosphate
Form Powder
InChI Key DUYSYHSSBDVJSM-KRWOKUGFSA-N
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(O)(O)=O
IUPAC Name {[(2S,3R,4E)-2-amino-3-hydroxyoctadec-4-en-1-yl]oxy}phosphonic acid
Molecular Weight (g/mol) 379.48
Show More Show Less
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only