Learn More
Eisco™ Spare Extractors for Soxhlet Apparatus
Sockets and cones feature ground glass. A variety of sizes available
Supplier:  Eisco™ CH0888M
 Afficher plus de versions de ce produit
Numéro de catalogue. S29518
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Spare extractor for Soxhlet apparatus
  • Variety of capacity, socket and cone sizes available
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • Spécifications
    Spécifications
    1
    1000 mL
    Distillation
    9.8 cm
    Socket size 55/44, Cone size 34/35
    Borosilicate Glass
    43 cm
    missing translation for 'recommendations'
    missing translation for 'recommendations'
    Videos
    FDS
    Documents
    Documents
    Promotions
    Promotions
    1 year limited manufacturer warranty