missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solid Neoprene Rubber Stoppers

Recommended where rubber would be attacked or deteriorated

$48.04 - $618.14

Specifications

Color Green
Material Neoprene
Product Type Solid Stopper
For Use With (Application) Resists petroleum products, fats, oils and organic chemicals
Length (Metric) 2.5 cm
View More Specs

Products 17
Catalog Number Size Stopper Number Price Quantity & Availability  
Catalog Number Size Stopper Number Price Quantity & Availability  
14141C
View Documents Promotion Details
2 2
Pack of 44 Each for $116.54
Case of 5 Pack for $582.70
Only null left
 
14141E
View Documents Promotion Details
4 4
Pack of 27 Each for $110.61
Case of 5 Pack for $553.05
Only null left
 
14141J
View Documents Promotion Details
7 7
Pack of 12 Each for $109.89
Case of 5 Pack for $549.45
Only null left
 
14141D
View Documents Promotion Details
3 3
Pack of 32 Each for $117.00
Case of 5 Pack for $585.00
Only null left
 
14141T
View Documents Promotion Details
13.5 13.5
Each for $48.04
Case of 13 Each for $624.52
Only null left
 
14141M
View Documents Promotion Details
10 10
Pack of 8 Each for $129.96
Case of 5 Pack for $649.80
Only null left
 
14141N
View Documents Promotion Details
10.5 10.5
Pack of 7 Each for $137.04
Case of 5 Pack for $685.20
Only null left
 
14141B
View Documents Promotion Details
1 1
Pack of 48 Each for $122.14
Case of 5 Pack for $610.70
Only null left
 
14141A
View Documents Promotion Details
0 0
Pack of 68 Each for $113.93
Case of 5 Pack for $569.65
Only null left
 
14141F
View Documents Promotion Details
5 5
Pack of 23 Each for $123.29
Case of 5 Pack for $616.45
Only null left
 
14141V
View Documents Promotion Details
14 14
Each for $72.71
Case of 8 Each for $581.68
Only null left
 
14141Q
View Documents Promotion Details
11.5 11.5
Pack of 4 Each for $133.64
Case of 5 Pack for $668.20
Only null left
 
14141K
View Documents Promotion Details
8 8
Pack of 10 Each for $116.54
Case of 5 Pack for $582.70
Only null left
 
14141W
View Documents Promotion Details
15 15
Case of 7 Each for $618.14
Only null left
 
14141R
View Documents Promotion Details
12 12
Pack of 5 Each for $160.79
Case of 5 Pack for $803.95
Only null left
 
14141P
View Documents Promotion Details
11 11
Pack of 6 Each for $126.57
Case of 5 Pack for $632.85
Only null left
 
14141L
View Documents Promotion Details
9 9
Pack of 8 Each for $129.14
Case of 5 Pack for $645.70
Only null left
 
Description

Description

Recommended where rubber would be attacked or deteriorated

  • Solid neoprene
  • Resists petroleum products, fats, oils and organic chemicals

The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).

Specifications

Specifications

Green
Solid Stopper
2.5 cm
Neoprene
Resists petroleum products, fats, oils and organic chemicals
Videos
SDS
Documents

Documents