Learn More
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
 Afficher plus de versions de ce produit
Numéro de catalogue. 14141P
Il en reste null
Description
Description
Recommended where rubber would be attacked or deteriorated
  • Solid neoprene
  • Resists petroleum products, fats, oils and organic chemicals
Spécifications
Identifiants chimiques
Green
Solid Stopper
2.5 cm
Neoprene
Resists petroleum products, fats, oils and organic chemicals
Spécifications
Solid Stopper
5.6 cm
11
Green
Neoprene
4.8 cm
2.5 cm
11
Resists petroleum products, fats, oils and organic chemicals
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS