Learn More
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
Description
Recommended where rubber would be attacked or deteriorated
- Solid neoprene
- Resists petroleum products, fats, oils and organic chemicals
Chemical Identifiers
| Green | |
| Solid Stopper | |
| 2.5 cm |
| Neoprene | |
| Resists petroleum products, fats, oils and organic chemicals |
Specifications
| Solid Stopper | |
| 4.5 cm | |
| 2.5 cm | |
| Green | |
| Resists petroleum products, fats, oils and organic chemicals |
| 3.7 cm | |
| 9 | |
| 9 | |
| Neoprene |