Learn More
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
 View more versions of this product
Catalog No. 14141K
Only null left
Description
Description
Recommended where rubber would be attacked or deteriorated
  • Solid neoprene
  • Resists petroleum products, fats, oils and organic chemicals
Specifications
Chemical Identifiers
Green
Solid Stopper
2.5 cm
Neoprene
Resists petroleum products, fats, oils and organic chemicals
Specifications
Solid Stopper
4.1 cm
2.5 cm
Resists petroleum products, fats, oils and organic chemicals
Neoprene
3.3 cm
8
8
Green
Product Suggestions
Product Suggestions