Learn More
Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
Description
Recommended where rubber would be attacked or deteriorated
- Solid neoprene
- Resists petroleum products, fats, oils and organic chemicals
Identifiants chimiques
| Green | |
| Solid Stopper | |
| 2.5 cm |
| Neoprene | |
| Resists petroleum products, fats, oils and organic chemicals |
Spécifications
| Solid Stopper | |
| 1.7 cm | |
| 2.5 cm | |
| Neoprene | |
| Resists petroleum products, fats, oils and organic chemicals |
| 1.3 cm | |
| 0 | |
| 0 | |
| Green |