Learn More
Sodium thiosulfate, 0.01N Standardized Solution
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
$80.49 - $252.44
Identifiants chimiques
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AA35643K2
|
Thermo Scientific Chemicals
035643K2 |
1 L |
chaque for $80.49
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA35643K7
|
Thermo Scientific Chemicals
035643K7 |
4 L |
chaque for $162.77
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AA35643M4
|
Thermo Scientific Chemicals
035643M4 |
20 L |
chaque for $252.44
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Sodium thiosulfate, 0.01N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane |
Spécifications
| 7772-98-7 | |
| Na2S2O3 | |
| 0.01N | |
| Immiscible with alcohol. | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.1 | |
| Liquid |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 24477 | |
| 1 L | |
| Sodium thiosulfate, Standardized Solution |
Sécurité et manipulation
EINECSNumber : 231-867-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only