missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Crystal, ACS, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1495125GM

 Afficher plus de versions de ce produit

Numéro de catalogue. 18613900


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

540-72-7
125 g
ACS
1
98%
Amber Glass Bottle

Identifiants chimiques

540-72-7
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.