missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Crystal, ACS, 98%, Spectrum™ Chemical

Catalog No. p-7166371
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Catalog No. Quantity Packaging
18613899 25 g Amber Glass Bottle
18613900 125 g Amber Glass Bottle
18613901 500 g Amber Glass Bottle
18613902 2.5 kg Amber Glass Bottle
18613903 12 kg Poly Pail
18613904 50 kg Fiber Drum
6 options

Catalog No. 18613899

Supplier: Spectrum Chemical Mfg Cor S149525GM

Only null left

Chemical Identifiers

CAS 540-72-7

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.