missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate Solution, Reagent Grade, Honeywell™

$524.37 - $524.37

Identifiants chimiques

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantité  
60-048-584
Afficher les documents
Honeywell Inc
3384433L
3 L Poly bottle
chaque for $524.37
Il en reste null
 
Description

Description

Spécifications

Identifiants chimiques

1344-09-8
212.14
ADPGKKZKGXANON-UHFFFAOYSA-N
23266
disodium;dioxido(oxo)silane
H10Na2O8Si
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
CHEBI:60720
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Spécifications

1344-09-8
H10Na2O8Si
MFCD00003492
sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O
212.14
CHEBI:60720
1.39g/mL at 25°C
3 L
Na2O(SiO2)x · xH2O
UN3266
ADPGKKZKGXANON-UHFFFAOYSA-N
disodium;dioxido(oxo)silane
23266
Poly bottle
Sodium silicate solution
Videos
Sécurité et manipulation

Sécurité et manipulation

P261-P280-P305 + P351 + P338-P310

missing translation for 'dotInformation' : Class 8/ PGIII

FDS
missing translation for 'documents'

missing translation for 'documents'