missing translation for 'onlineSavingsMsg'
Learn More

Sodium Silicate Solution, Reagent Grade, Solstice

Catalog No. 60048584
Change view
Click to view available options
Quantity:
3 L
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048584 3 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048584 Supplier Honeywell Inc Supplier No. 3384433L

Chemical Identifiers

CAS 1344-09-8
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
MDL Number MFCD00003492
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate, sodium silicate, waterglass, water glass, sodium siliconate, sodium polysilicate, sodium sesquisilicate, sodium silicate glass, sodium silicate solution, sodium water glass
PubChem CID 23266
ChEBI CHEBI:60720
IUPAC Name disodium;dioxido(oxo)silane
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Quantity 3 L
Linear Formula Na2O(SiO2)x · xH2O
UN Number UN3266
Packaging Poly bottle
Density 1.39g/mL at 25°C
Chemical Name or Material Sodium silicate solution
Hazard Statement P261-P280-P305 + P351 + P338-P310
DOTInformation Class 8/ PGIII