missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite, 5% Available Chlorine, Solution, Reagent, Spectrum™ Chemical
S1315, 7681-52-9, NaOCl
Supplier: Spectrum Chemical Mfg Cor S13154LT
Description
Spectrum™ Chemical Sodium Hypochlorite, 5 Percent Available Chlorine, Solution, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. The primary use of Sodium Hypochlorite in labs and manufacturing is in surface purification, textile bleaching, odor removal in water treatment facilities, and to detoxify cyanide baths in the metal processing industry.Spécifications
| 7681-52-9,7732-18-5,1310-73-2 | |
| 4 L | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| sodium hypochlorite | |
| Poly Bottle |
| 0.07,0.92,0.01 | |
| ClNaO | |
| [Na+].[O-]Cl | |
| 74.44 | |
| 1.07 |
Identifiants chimiques
| 7681-52-9 | |
| 74.44 | |
| sodium hypochlorite |
| ClNaO | |
| SUKJFIGYRHOWBL-UHFFFAOYSA-N | |
| [Na+].[O-]Cl |