missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Solution, Reagent, Spectrum™ Chemical

Catalog No. p-7158231
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
4 x 4 L
200 L
Packaging:
Poly Bottle
Poly Drum
Catalog No. Quantity Packaging
18606587 6 x 500 mL Poly Bottle
18606588 4 L Poly Bottle
18606589 4 x 4 L Poly Bottle
18606590 200 L Poly Drum
4 options

Catalog No. 18606587

Supplier: Spectrum Chemical Mfg Cor S1315500MLPLCS6

Only null left

S1315, 7681-52-9, NaOCl

Spectrum™ Chemical Sodium Hypochlorite, 5 Percent Available Chlorine, Solution, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. The primary use of Sodium Hypochlorite in labs and manufacturing is in surface purification, textile bleaching, odor removal in water treatment facilities, and to detoxify cyanide baths in the metal processing industry.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5 , 1310-73-2
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl

Specifications

CAS Max % 0.07, 0.92, 0.01
Quantity 6 x 500 mL
Packaging Poly Bottle
Specific Gravity 1.07