missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, 1.0M, Honeywell Fluka™
Volumetric, 1.0Â M NaOH (1.0N)
Supplier: Honeywell Chemicals 3195116X1L
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Spécifications
| Sodium Hydroxide Solution | |
| HNaO | |
| MFCD00003548 | |
| 1824 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| sodium;hydroxide | |
| 14798 | |
| 40g/mol | |
| 3mmHg (37°C) |
| 1310-73-2 | |
| NaOH | |
| 1.0M | |
| 1.04g/mL at 25°C | |
| 6 x 1 L | |
| [OH-].[Na+] | |
| 39.997 | |
| CHEBI:32145 | |
| <1 (vs air) |
Identifiants chimiques
| 1310-73-2 | |
| 39.997 | |
| HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
| 14798 | |
| sodium;hydroxide |
| HNaO | |
| MFCD00003548 | |
| sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
| CHEBI:32145 | |
| [OH-].[Na+] |
Sécurité et manipulation
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H314