missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 1.0M, Honeywell Fluka™

Numéro de catalogue. p-7072057
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
6001451 500 mL
6001449 1 L
6001452 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. 6001451 Fournisseur Honeywell Chemicals Code fournisseur 319511500ML
Il en reste null

Volumetric, 1.0 M NaOH (1.0N)

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Identifiants chimiques

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Spécifications

Linear Formula NaOH
Concentration 1.0M
UN Number 1824
Density 1.04g/mL at 25°C
Quantity 500 mL
Formula Weight 40g/mol
Vapor Density <1 (vs air)
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium Hydroxide Solution
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314