Learn More
Metro™ SmartWall Wire Grid
Metro SmartWall Wire Grids create storage and workstation systems for underutilized wall space.
Supplier:  Metro™ WG1848K4
 Afficher plus de versions de ce produit
Numéro de catalogue. 13460386
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Grids may attach directly to SmartWall uprights.
  • No tools are necessary.
  • Grids may not overhang the uprights by more than 6 in. (152 mm).
  • Grid openings measure approximately 3 x 3 in. (76 x 76 mm).
Spécifications
Spécifications
Product will be free from defects in material and workmanship under normal use, service and maintenance for a period of one (1) year from the date of shipment of the Product by Seller, unless otherwise specified by Seller in writing.
0.5 in.
457.2 mm
48 in.
10.8 lb.
1219.2 mm
Not Autoclavable
Epoxy-coated Steel
Wire Grid
All Laboratory and Clean Room Non-Autoclavable Applications
18 in.
4.8 kg
12.7 mm
SmartWall Shelving
Gray
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS