Learn More
Thermo Scientific Chemicals Simethicone
CAS: 8050-81-5 | C6H18O4Si3 | 238.46 g/mol
$233.51 - $710.83
Identifiants chimiques
| CAS | 8050-81-5 |
|---|---|
| Molecular Formula | C6H18O4Si3 |
| Molecular Weight (g/mol) | 238.46 |
| InChI Key | AMTWCFIAVKBGOD-UHFFFAOYSA-N |
| IUPAC Name | 3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione |
| SMILES | O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
AC465170100
|
Thermo Scientific Chemicals
465170100 |
10 g |
chaque for $233.51
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
AC465170500
|
Thermo Scientific Chemicals
465170500 |
50 g |
chaque for $710.83
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 8050-81-5 | |
| 238.46 | |
| 3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione |
| C6H18O4Si3 | |
| AMTWCFIAVKBGOD-UHFFFAOYSA-N | |
| O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C |
Spécifications
| 8050-81-5 | |
| 10 g | |
| AMTWCFIAVKBGOD-UHFFFAOYSA-N | |
| 3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione | |
| Simethicone |
| C6H18O4Si3 | |
| Solubility aq. soln.: .. Other solubilities: soluble in chloroform | |
| O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C | |
| 238.46 |
Sécurité et manipulation
GHS H Statement Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
GHS Signal Word: Warning
RUO – Research Use Only