missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Simethicone

CAS: 8050-81-5 | C6H18O4Si3 | 238.46 g/mol

Supplier:  Thermo Scientific Chemicals 465170100

 View more versions of this product

Catalog No. AC465170100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Simethicone
C6H18O4Si3
Solubility aq. soln.: .. Other solubilities: soluble in chloroform
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
238.46
8050-81-5
10 g
AMTWCFIAVKBGOD-UHFFFAOYSA-N
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione

Chemical Identifiers

8050-81-5
238.46
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
C6H18O4Si3
AMTWCFIAVKBGOD-UHFFFAOYSA-N
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only