missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self-Expiring Parking Permit

Control parking with an economical and effective solution. Brady™ Moderate Weather Self-Expiring Parking Permits are a durable solution that make parking control easy.

$291.89 - $291.89

Spécifications

Length (Metric) 5 in.
Color Red on white
Dimensions (L x W) 5 in. x 3 in.
Width (English) 3 in.
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Product Type For Use With (Application) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Product Type For Use With (Application) Prix Quantité  
19-103-180
Afficher les documents
Brady™
95681
Moderate weather (50-80°F/10-25°C) Temporary
paquet de 500 chaque for $291.89
Il en reste null
 
Description

Description

  • Temporary parking permits expire during the day by progressively revealing the letters E-X-P-I-R-E-D
  • Expiration process starts once the self-adhesive cover is applied
Spécifications

Spécifications

5 in.
Red on white
5 in. x 3 in.
3 in.
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'