missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Self-Expiring Parking Permit

Catalog No. 19103180
Change view
Click to view available options
Product Type:
Moderate weather (50-80°F/10-25°C)
For Use With (Application):
Temporary
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
19103180 Moderate weather (50-80°F/10-25°C) Temporary
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19103180 Supplier Brady™ Supplier No. 95681

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Control parking with an economical and effective solution. Brady™ Moderate Weather Self-Expiring Parking Permits are a durable solution that make parking control easy.

  • Temporary parking permits expire during the day by progressively revealing the letters E-X-P-I-R-E-D
  • Expiration process starts once the self-adhesive cover is applied

Specifications

Length (Metric) 5 in.
Color Red on white
Dimensions (L x W) 5 in. x 3 in.
For Use With (Application) Temporary
Width (English) 3 in.
Product Type Moderate weather (50-80°F/10-25°C)