missing translation for 'onlineSavingsMsg'
Learn More

Agilent Technologies™ Seahorse XF DMEM Medium Assay Kit

Catalog No. 103680100
Catalog No. 103680100 Supplier Agilent Technologies™ Supplier No. 103680100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seahorse XF DMEM Medium Assay Kit includes XF DMEM medium with a pH of 7.4, XF 1.0 M Glucose Solution, XF 100 mM Pyruvate Solution, and XF 200 mM Glutamine Solution.

  • Specifically formulated for Agilent™ Seahorse XF assays to achieve optimal results
  • Contains no glucose, glutamine, or pyruvate, allowing for full customization
  • Offers options of with or without phenol red, DMEM- or RPMI-based
  • XF media with pH 7.4 is also available, simplifying the workflow, saving time for assay preparation, and ensuring consistent results across experiments
  • Includes: XF DMEM medium pH 7.4 (500 mL), XF 1.0 M Glucose Solution (50 mL), XF 100 mM Pyruvate Solution (50 mL), and XF 200 mM Glutamine Solution (50 mL)

Specifications

Classification Animal Origin-free
Concentration 1X
Form Liquid
Product Type Seahorse XF Assay
Serum Level Serum-free
Sterility Sterile
With Additives None
Without Additives None
Formulation XF Assay with Supplements
Culture Type Adherent Cell Culture