Learn More
Thermo Scientific™ SAF Fixative Single Vial
Collect, preserve, concentrate and permanently stain intestinal parasites with a quality fecal transport which comes with industry standard color-coding and sporked caps.
Supplier:  Thermo Scientific™ RE21921
 View more versions of this product
Catalog No. R21921
Only null left
Description
Description
Collect, preserve, concentrate and permanently stain intestinal parasites with Thermo Scientfic™ SAF Fixative Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing. Look to us for convenient kit packaging options, including bulk kits. Multilingual patient collection instructions are included. Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
SAF Fixative
15 mL
120 Vials/Pk.
SAF Fixative Single Vial
Single vial. Packaged in partitioned boxes.
Product Suggestions
Product Suggestions