missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G

Suitable for fluorescent labelling | CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

$66.51 - $174.10

Identifiants chimiques

CAS 989-38-8
Molecular Formula C28H31ClN2O3
Molecular Weight (g/mol) 479.017
MDL Number MFCD00012665
InChI Key VYXSBFYARXAAKO-UHFFFAOYSA-N
Synonym Basic Red 1; C.I. 45160
PubChem CID 51358423
IUPAC Name ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
SMILES [H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAJ6231514
Afficher les documents
Thermo Scientific Chemicals
J6231514
25 g
chaque for $66.51
Il en reste null
 
AAJ6231522
Afficher les documents
Thermo Scientific Chemicals
J6231522
100 g
chaque for $174.10
Il en reste null
 
Description

Description

Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
Spécifications

Identifiants chimiques

989-38-8
479.017
VYXSBFYARXAAKO-UHFFFAOYSA-N
51358423
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
C28H31ClN2O3
MFCD00012665
Basic Red 1; C.I. 45160
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride

Spécifications

989-38-8
MFCD00012665
Basic Red 1; C.I. 45160
VYXSBFYARXAAKO-UHFFFAOYSA-N
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
51358423
Brown to Red
Powder
C28H31ClN2O3
3900071
Soluble in water.
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
479.017
479.02
25 g
Rhodamine 6G
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-584-9

RTECSNumber : DH0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

RUO – Research Use Only