missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rhodamine 6G

Suitable for fluorescent labelling | CAS: 989-38-8 | C28H31ClN2O3 | 479.017 g/mol

Supplier:  Thermo Scientific Chemicals J6231522

 View more versions of this product

Catalog No. AAJ6231522


Only null left

Description

Description

Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodamine dyes are used extensively in biotechnology applications such as fluorescence microscopy, flow cytometry,fluorescence correlation spectroscopy and ELISA.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
Specifications

Specifications

Rhodamine 6G
C28H31ClN2O3
3900071
Soluble in water.
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
479.017
479.02
100 g
989-38-8
MFCD00012665
Basic Red 1; C.I. 45160
VYXSBFYARXAAKO-UHFFFAOYSA-N
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
51358423
Brown to Red
Powder

Chemical Identifiers

989-38-8
479.017
VYXSBFYARXAAKO-UHFFFAOYSA-N
51358423
[H+].CCNC1=C(C=C2C(=C1)OC3=CC(=NCC)C(=CC3=C2C4=CC=CC=C4C(=O)OCC)C)C.[Cl-]
C28H31ClN2O3
MFCD00012665
Basic Red 1; C.I. 45160
ethyl 2-[3-(ethylamino)-6-ethylimino-2,7-dimethylxanthen-9-yl]benzoate;hydron;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-584-9

RTECSNumber : DH0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only