Learn More
Restek™ Mininert Precision Sampling Valves for Vials DFS Item
Convenient valves for repetitive sampling and limiting content exposure to the silicon septum.
Supplier:  Restek™ RK24901
 Afficher plus de versions de ce produit
Numéro de catalogue. 06719051
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Available for both screw-cap and crimp-top vials
  • Crimp-top valve for 20mm ID glassware slides into the neck of the vial
  • Turn the threaded flange to secure a tight fit
Spécifications
Spécifications
Valve
12 Pack
Screw-Thread Vials
Precision Sampling
15 mm/425
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS