Learn More
Restek™ Mininert Precision Sampling Valves for Vials DFS Item
Convenient valves for repetitive sampling and limiting content exposure to the silicon septum.
$529.43 - $725.32
Specifications
Type Valve
For Use With (Application) Precision Sampling
Packaging Quantity 12 Pack
View More Specs
Products 6
Catalog Number Mfr. No. For Use With Thread Size Price Quantity  
Catalog Number Mfr. No. For Use With Thread Size Price Quantity  
06719050 DFS Item
View Documents
Restek™
RK24900
Screw-Thread Vials 13 mm/425
Pack of 12 Each for $535.92
Only null left
06719052 DFS Item
View Documents
Restek™
RK24902
Screw-Thread Vials 18 mm/400
Pack of 12 Each for $575.38
Only null left
06719053 DFS Item
View Documents
Restek™
RK24903
Screw-Thread Vials 20 mm/400
Pack of 12 Each for $611.09
Only null left
06719055 DFS Item
View Documents
Restek™
RK24905
Crimp-Top Vial 20 mm
Pack of 12 Each for $725.32
Only null left
06719054 DFS Item
View Documents
Restek™
RK24904
Screw-Thread Vials 24 mm/400
Pack of 12 Each for $588.43
Only null left
06719051 DFS Item
View Documents
Restek™
RK24901
Screw-Thread Vials 15 mm/425
Pack of 12 Each for $529.43
Only null left
Description
Description
  • Available for both screw-cap and crimp-top vials
  • Crimp-top valve for 20mm ID glassware slides into the neck of the vial
  • Turn the threaded flange to secure a tight fit
Specifications
Specifications
Valve
Precision Sampling
12 Pack
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'