Learn More
Invitrogen™ Human Renin ELISA Kit
ELISA
$908.00 - $6660.00
Spécifications
| Accession Number | P00797 |
|---|---|
| Assay Range | 93.8 to 6000 pg/mL |
| Assay Sensitivity | 32.6 pg/ml |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
BMS2271
|
Invitrogen™
BMS2271 |
96 Tests |
chaque for $908.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
BMS2271TEN
|
Invitrogen™
BMS2271TEN |
10 x 96 Tests |
chaque for $6,660.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
The Renin Human ELISA Kit quantitates Hu Renin in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Renin.
Principle of the method
The Human Renin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Renin is a highly specific endopeptidase, whose only known function is to generate angiotensin I from angiotensinogen in the plasma, initiating a cascade of reactions that produce an elevation of blood pressure and increased sodium retention by the kidney.Spécifications
| P00797 | |
| 32.6 pg/ml | |
| ELISA | |
| Human | |
| 5972 | |
| 5.1% | |
| Pre-coated 96 well plate, Standard (2 vials), Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, Streptavidin-HRP, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 93.8 to 6000 pg/mL | |
| HRP | |
| Cell Lysates, Plasma, Serum | |
| Colorimetric Microplate Reader | |
| REN | |
| 3.9 | |
| HRP | |
| Cell Lysate 50 μL, Plasma 50 μL, Serum 50 μL | |
| Renin | |
| 3 hrs 30 mins |