Learn More
Applied Biosystems™ RapidINTEL™ Positive Control Cartridge
The Applied Biosystems RapidINTEL Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance.
Supplier:  Applied Biosystems™ P044396
Numéro de catalogue. P044396
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
The Applied Biosystems RapidINTEL Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single-use/disposable.
Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Spécifications
Spécifications
Cartridge
RapidHIT™ ID System
Human Identification
1 Cartridge
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS