Learn More
Azure Biosystems Radiance Q
Quantitative HRP substrate for chemiluminescent Western blots
Supplier:  Azure Biosystems AC2101
Numéro de catalogue. AC2101AZ
Il en reste null
Description
Description
  • Radiance Q chemiluminescent substrate is specially formulated for use with CCD imagers. This novel horseradish peroxidase (HRP) substrate provides a strong, long-lasting signal, the broadest useful linear range, and high sensitivity for the most quantitative chemiluminescent Western assays.
  • Size: 150 mL
Spécifications
Spécifications
ECL Western Blot Detection Reagent
Liquid
150 mL
Horseradish Peroxidase
Long Lasting
Chemiluminescence
Western Blot
HRP
Chemiluminescent
1500 cm2
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS