missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-Phenylcyclohexanone 97.0+%, TCI America™
Supplier: TCI America P14991G
Spécifications
| (R)-3-Phenylcyclohexanone | |
| Yellow | |
| MFCD08276410 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one | |
| 11041212 | |
| ≥97.0% (GC) |
| 34993-51-6 | |
| C12H14O | |
| 1 g | |
| O=C1CCCC(C1)C1=CC=CC=C1 | |
| 174.24 | |
| 174.24 | |
| Liquid |
Identifiants chimiques
| 34993-51-6 | |
| 174.24 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one |
| C12H14O | |
| MFCD08276410 | |
| 11041212 | |
| O=C1CCCC(C1)C1=CC=CC=C1 |
Sécurité et manipulation
missing translation for 'tsca' : No
Recommended Storage : Refrigerator