Learn More
Corning™ PYREX™ Reusable Media Storage Bottles
GREENER_CHOICE
Ideal for media, buffers, aqueous and nonaqueous solutions.
$101.00 - $726.93
Spécifications
Material Borosilicate Glass
Closure Color Orange
Shape Cylindrical
Closure Material Polypropylene
voir les spécifications
Avertissement:
Bottles should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.
Produits 12
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Mouth Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Mouth Quantity Prix Quantity & Availability  
06423A
Accessoires Afficher les documents Détails de la promotion
Corning™
139525
25 mL Narrow 10/Cs.
caisse de 10 chaque for $306.15
Il en reste null
06423B
Accessoires Afficher les documents Détails de la promotion
Corning™
139550
50 mL Narrow 10/Cs.
caisse de 10 chaque for $237.41
Il en reste null
064141A
Afficher les documents Détails de la promotion
Corning™
1395100
100 mL Narrow 10/Cs.
caisse de 10 chaque for $239.74
Il en reste null
10462711
Afficher les documents Détails de la promotion
Corning™
1395150
150 mL Wide 10/Cs.
caisse de 10 chaque for $254.17
Il en reste null
064141B
Accessoires Afficher les documents Détails de la promotion
Corning™
1395250
250 mL Wide 10/Cs.
caisse de 10 chaque for $276.70
Only null left
Edge
064141C
View Documents Promotion Details
Corning™
1395500
500 mL Wide 10/Cs.
caisse de 10 chaque for $322.59
Only null left
Edge
10462712
View Documents Promotion Details
Corning™
1395750
750 mL Wide 10/Cs.
caisse de 10 chaque for $336.91
Only null left
064141D
View Documents Promotion Details
Corning™
13951L
1 L Wide 10/Cs.
caisse de 10 chaque for $396.41
Only null left
064141E
Accessories View Documents Promotion Details
Corning™
13952L
2000 mL Wide 1/Cs.
chaque for $101.00
caisse de 10 chaque for $1,010.00
Only null left
Edge
10462710
View Documents Promotion Details
Corning™
13953X
3500 mL Narrow 1/Cs.
caisse for $333.13
Only null left
064141F
View Documents Promotion Details
Corning™
13955L
5000 mL Wide 1/Cs.
caisse for $378.43
Only null left
064141G
GREENER_CHOICE
View Documents Promotion Details
Corning™
139510L
10 L Narrow 1/Cs.
caisse for $726.93
Only null left
Description
Description
Ideal for media, buffers, aqueous and nonaqueous solutions
Heavy duty bottle which can be used for storage as well as mixing and sampling. Includes linerless, one-piece autoclavable polypropylene plug seal cap with drip free pouring rings. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Glass bead indicates full capacity line on 100 mL through 2L size. Bottles have permanent white enamel graduations and marking spots. Color-coded polypropylene caps are available in a choice of orange, green, purple, red, and light gray.
Caution: Bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT have caps tightened immediately after autoclaving, as the vacuum resulting from cooling can cause breakage. A pouring ring is not included on 25 mL size. A vented cap is NOT recommended for use on the 5L and 10L 1395 series bottle.
Order Info
Optional red melamine caps and PTFE rings are available for applications requiring temperatures to 180°C (356°F).
Specifications
Specifications
Borosilicate Glass
Orange
Cylindrical
Polypropylene