missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Reusable Media Storage Bottles

Catalog No. 10462712
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
750 mL
1 L
2000 mL
3500 mL
5000 mL
10 L
Mouth:
Narrow
Wide
Quantity:
1/Cs.
10/Cs.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Mouth Quantity
10462712 750 mL Wide 10/Cs.
06423A 25 mL Narrow 10/Cs.
06423B 50 mL Narrow 10/Cs.
064141A 100 mL Narrow 10/Cs.
10462711 150 mL Wide 10/Cs.
064141B 250 mL Wide 10/Cs.
064141C 500 mL Wide 10/Cs.
064141D 1 L Wide 10/Cs.
064141E 2000 mL Wide 1/Cs.
10462710 3500 mL Narrow 1/Cs.
064141F 5000 mL Wide 1/Cs.
064141G 10 L Narrow 1/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Bottles should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.
Catalog No. 10462712 Supplier Corning™ Supplier No. 1395750
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bottles should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.

Ideal for media, buffers, aqueous and nonaqueous solutions.

Ideal for media, buffers, aqueous and nonaqueous solutions

Heavy duty bottle which can be used for storage as well as mixing and sampling. Includes linerless, one-piece autoclavable polypropylene plug seal cap with drip free pouring rings. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Glass bead indicates full capacity line on 100 mL through 2L size. Bottles have permanent white enamel graduations and marking spots. Color-coded polypropylene caps are available in a choice of orange, green, purple, red, and light gray.

Caution: Bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT have caps tightened immediately after autoclaving, as the vacuum resulting from cooling can cause breakage. A pouring ring is not included on 25 mL size. A vented cap is NOT recommended for use on the 5L and 10L 1395 series bottle.

Order Info

Optional red melamine caps and PTFE rings are available for applications requiring temperatures to 180°C (356°F).

Specifications

Capacity 25.36 oz., 750 mL
Capacity (Metric) 750 mL
Material Borosilicate Glass
No. per Case 10/Cs.
No. per Pack 10/Pk.
Quantity 10/Cs.
Sterility Non-sterile
Closure Color Orange
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Material Polypropylene
Closure Size GL 45
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 25.36 oz.
Shape Cylindrical
Disposable Reusable
Product Type Media Storage Bottle
Show More Show Less