missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GVS PVDF-Plus™ Transfer Membrane
Improves detection limits for more sensitive Western blots.
Supplier: GVS 1214842
Description
- Superior strength: Can withstand aggressive handling or be used with automated equipment without breaking or tearing
- Low extractables: Ensures tests will be clean with consistent results
- Exceptional sensitivity: Detects low-level components
- Hydrophobic:For high protein binding
- Lot-to-lot consistency: Quality checks ensure consistent binding for dependable results every time
- BSA protein binding capacity is 125mg/cm2
- High range of chemical: Resistant to most commonly used chemicals compatible with chemically aggressive solvents
Identifiants chimiques
| 3 m | |
| Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses | |
| White | |
| PVDF |
| 15 cm | |
| 125 mg/cm2 | |
| Roll |
Spécifications
| 3 m | |
| Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses | |
| White | |
| PVDF |
| 15 cm | |
| 125 mg/cm2 | |
| Roll | |
| 0.22 μm |