missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GVS PVDF-Plus™ Transfer Membrane
Improves detection limits for more sensitive Western blots.
$575.97 - $625.72
Specifications
| Length (Metric) | 3 m |
|---|---|
| Width (Metric) | 15 cm |
| For Use With (Application) | Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses |
| Binding Capacity | 125 mg/cm2 |
| Color | White |
| Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Pore Size | Price | Quantity & Availability | |||||
|
0973250
|
GVS
1214842 |
0.22 μm |
Pack for $575.97
|
|
|||||
|
PV4HYB0010
|
GVS
1212781 |
0.45 μm |
Each for $625.72
|
|
|||||
Description
- Superior strength: Can withstand aggressive handling or be used with automated equipment without breaking or tearing
- Low extractables: Ensures tests will be clean with consistent results
- Exceptional sensitivity: Detects low-level components
- Hydrophobic:For high protein binding
- Lot-to-lot consistency: Quality checks ensure consistent binding for dependable results every time
- BSA protein binding capacity is 125mg/cm2
- High range of chemical: Resistant to most commonly used chemicals compatible with chemically aggressive solvents
Specifications
| 3 m | |
| Western blots, Immunoblotting, Solid phase assays, Amino acid or protein analyses | |
| White | |
| PVDF |
| 15 cm | |
| 125 mg/cm2 | |
| Roll |