missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Protector™Acid Storage Cabinets
Support Labconco fume hoods and safely store and vent acids and other corrosive liquids
Supplier: Labconco™ 9905200
Comprend:
Vacuum-formed PVC liner tray, two rear vent connection cutouts for venting the cabinet into a fume hood, four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.
Description
- Corrosion-resistant polyethylene-lined interior
- Manual-closing, non-locking door(s)
Spécifications
| Storage Cabinet | |
| Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
| 24 in. | |
| Single Left Hinged Door | |
| 55.88 cm | |
| 31.5 to 32.75 in. | |
| 800 lb. | |
| Light Gray |
| 22 in. | |
| Vacuum formed PVC liner tray, 2 rear vent connection cut outs for venting the cabinet into a fume hood, 4 leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30in. | |
| 54.43 kg | |
| 120 lb. | |
| 362.8 kg | |
| ADA | |
| 80 to 83.18 cm | |
| Steel |