missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 50 Coveralls with Hood
Description
Microporous film coveralls provide protection from non-hazardous aerosols, light liquid splash, and dry particles.
- Made with a microporous film that is laminated to a spundbonded polypropylene substrate
- Front zipper closure
- Elastic wrists, ankles
- Attached hood
- Serged seams
- Collar
- White
- Thickness: 10 mils
Spécifications
Spécifications
| Product Type | General Purpose Coveralls |
| Size | Small |
| Gender | Unisex |
| Material | ProShield |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Hood Type | Standard, Elastic Face |
| Afficher plus de résultats |