Learn More
Promega Restriction Enzymes - Pst I Blue/White Cloning Qualified (H)
Capable of digesting DNA in 15 minutes or less. Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications. (C TGCA/G G\ACGT C)
Supplier: Promega R6111
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, provides a higher level of quality control for enzymes used in cloning applications
Spécifications
| 10 U/μL | |
| 37°C | |
| Provides a higher level of quality control for Enzymes used in cloning applications | |
| -30°C to -10°C | |
| CTGCA.G G.ACGTC | |
| Liquid |
| 1 x 1mL 10X Buffer H, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 3000U PstI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
| 7.3 | |
| 10mM Tris HCl (pH 7.3), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.15% Triton™ X-100, 0.5mg/mL BSA, 50% Glycerol | |
| 3000 U | |
| PstI |