missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega P450-Glo™ CYP450 Assay Systems
Provide a complete set of reagents for performing luminescent cytochrome P450 assays
Supplier: Promega V8891
Comprend: Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
Description
- A Luminescent Method and Screening System for Measuring Activity
- Screening system includes complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
Identifiants chimiques
| -30°C to -10°C | |
| Homogeneous, Luminescence |
Spécifications
| 10 mL | |
| Homogeneous, Luminescence | |
| 1 x 1 Each Luciferin detection reagent, 1 x 10mL Reconstitution Buffer with Esterase, 1 x 240μg Luciferin-ME EGE |
| -30°C to -10°C | |
| For measuring cytochrome P450 activity | |
| P450-Glo CYP2D6 Screening Assay |
- U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.