missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega P450-Glo™ CYP450 Assay Systems
Description
Includes
Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
- A Luminescent Method and Screening System for Measuring Activity
- Screening system includes complete set of reagents
- Luminescent format eliminates need for time-consuming analyses
- Broad dynamic range and low background
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer, 1 x 1mL Luciferin-H 5mM -30°C to -10°C |
| Detection Method | Homogeneous, Luminescence |
| For Use With (Application) | For measuring cytochrome P450 activity |
| Product Type | P450-Glo CYP2C9 Assay |
- U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
- The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.