Learn More
Promega P450-Glo™ CYP450 Assay Systems
Provide a complete set of reagents for performing luminescent cytochrome P450 assays
Supplier:  Promega V8752
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. PRV8752
Il en reste null
Comprend: Luciferin Detection Reagent; P450-Glo Buffer; 5mM Luciferin-CEE, -ME, -H, -BE, -PFBE, -PPXE, or IPA
Description
Description
  • A Luminescent Method and Screening System for Measuring Activity
  • Screening system includes complete set of reagents
  • Luminescent format eliminates need for time-consuming analyses
  • Broad dynamic range and low background
Spécifications
Identifiants chimiques
-30°C to -10°C
Homogeneous, Luminescence
Spécifications
50 mL
Homogeneous, Luminescence
1 x 1 Each Luciferin detection reagent, 1 x 50mL Reconstitution Buffer, 1 x 350μL Luciferin-CEE 5mM
-30°C to -10°C
For measuring cytochrome P450 activity
P450-Glo CYP1A1 Assay
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS
Documents
Documents
Promotions
Promotions
  • U.S. Patent 6,602,677; Australian Pat. No. 754312 and other patents pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673.