missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Prepared Microscope Slide - Stomach Composite Section From Mammal
Prepared microscope composite slide of mammalian stomach
Supplier: Eisco™ BS18009
Description
- Pre-cleaned glass slide with ground edges
- Professionally stained samples for best visualization
- Ideal for biology classrooms to explore structure-function relationships as per NGSS standards
- Shows general structures and tissues of mammalian stomach
Spécifications
| 1 | |
| Glass | |
| Microscopy | |
| Formalin | |
| 1 | |
| 1 x 3 in. |
| 10% | |
| Microbiology Slide | |
| China | |
| Cardboard Mailer | |
| Dog | |
| Prepared Slide |