missing translation for 'onlineSavingsMsg'
Learn More

PPP1CA Polyclonal Antibody, Bethyl Laboratories DFS Item

Rabbit Polyclonal Antibody

$344.88 - $344.88

Spécifications

Antigen PPP1CA
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
501558751 DFS Item
Afficher les documents
Bethyl Laboratories Inc
A300904AT
20 μL
chaque for $344.88
Il en reste null
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to

The protein encoded by this gene is one of the three catalytic subunits of protein phosphatase 1 (PP1). PP1 is a serine/threonine specific protein phosphatase known to be involved in the regulation of a variety of cellular processes, such as cell division, glycogen metabolism, muscle contractility, protein synthesis, and HIV-1 viral transcription. Increased PP1 activity has been observed in the end stage of heart failure. Studies in both human and mice suggest that PP1 is an important regulator of cardiac function. Mouse studies also suggest that PP1 functions as a suppressor of learning and memory. Three alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécifications

Spécifications

PPP1CA
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
PPP1CA, PPP1A
Between 280 and C-term
Primary
4° C
PPP1CA
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse
P62136, P62137
19045, 5499
IgG
Antigen affinity chromatography
Antibody
Videos
FDS
Documents

Documents