missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Dibasic Trihydrate, ≥99%, MP Biomedicals
Supplier: MP Biomedicals Inc 0219484590
Spécifications
| Potassium Phosphate Dibasic Trihydrate | |
| 500 g | |
| H7K2O7P | |
| dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
| O.O.O.[K+].[K+].OP([O-])([O-])=O | |
| 228.22 | |
| 228.2 |
| 16788-57-1 | |
| ≥99% | |
| MFCD00149926 | |
| XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
| dipotassium trihydrate hydrogen phosphate | |
| 16217523 |
Identifiants chimiques
| 16788-57-1 | |
| 228.22 | |
| XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
| 16217523 | |
| O.O.O.[K+].[K+].OP([O-])([O-])=O |
| H7K2O7P | |
| MFCD00149926 | |
| dipotassium hydrogen phosphate trihydrate, dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
| dipotassium trihydrate hydrogen phosphate |
Sécurité et manipulation
Recommended Storage : Room Temperature